missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)naphthalene, 95%

Product Code. 10627062 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627062 500 g Glass bottle 500g
10365930 5 g Glass bottle 5g
10375930 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10627062 Supplier Thermo Scientific Acros Supplier No. 109455000
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86-52-2 | C11H9Cl | 176.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-52-2
Molecular Formula C11H9Cl
Molecular Weight (g/mol) 176.65
MDL Number MFCD00004042
InChI Key XMWGTKZEDLCVIG-UHFFFAOYSA-N
Synonym 1-chloromethyl naphthalene, naphthalene, 1-chloromethyl, 1-naphthylmethyl chloride, 1-menaphthyl chloride, 1-chlormethyl naftalen, chloromethyl naphthalene, 1-chloro methylnaphthalene, alpha-naphthylmethyl chloride
PubChem CID 6845
IUPAC Name 1-(chloromethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 31°C to 31°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 291°C to 292°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range Remainder 2-chloromethylnaphtalene (HPLC)
Packaging Glass bottle
Linear Formula C10H7CH2Cl
Refractive Index 1.6350 to 1.639
Quantity 500 g
Beilstein 05, 566
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 176.65
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1-(Chloromethyl)naphthalene, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(Chloromethyl)naphthalene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.