missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)naphthalene, TRC

Product Code. 30350343
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350343 10 g -
30349894 5 g 5g
30354573 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350343 Supplier Toronto Research Chemicals Supplier No. TRCC36803010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-52-2
Molecular Formula C11H9Cl
Molecular Weight (g/mol) 176.64
Synonym 1-Naphthalenylmethyl Chloride, 1-Naphthalylmethyl Chloride, 1-Naphthylmethyl Chloride, NSC 8473, α-(Chloromethyl)naphthalene, α-Naphthylmethyl Chloride
IUPAC Name 1-(chloromethyl)naphthalene
SMILES ClCC1=CC=CC2=CC=CC=C12

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H9Cl/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2
Formula Weight 176.0393
Percent Purity >95%
Chemical Name or Material 1-(Chloromethyl)naphthalene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.