missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorooctane, 99%

Product Code. 11376906
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11376906

Brand: Thermo Scientific Alfa Aesar L04358.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-85-3 | C8H17Cl | 148.674 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-85-3
Molecular Formula C8H17Cl
Molecular Weight (g/mol) 148.674
MDL Number MFCD00001022
InChI Key CNDHHGUSRIZDSL-UHFFFAOYSA-N
Synonym n-octyl chloride, octyl chloride, octane, 1-chloro, capryl chloride, chlorooctane, octane, chloro, 1-octyl chloride, dsstox_cid_1543, dsstox_rid_76201, dsstox_gsid_21543
PubChem CID 8142
IUPAC Name 1-chlorooctane
SMILES CCCCCCCCCl

Specifications

Melting Point -58°C
Density 0.873
Boiling Point 182°C to 183°C
Flash Point 63°C (145°F)
Refractive Index 1.4298
Quantity 500 mL
UN Number UN3082
Beilstein 1697464
Formula Weight 148.68
Percent Purity 99%
Chemical Name or Material 1-Chlorooctane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.