missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropentane, 98%, ACROS Organics™

Product Code. 10269773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269773 500 mL Glass bottle 500mL
10052880 5 mL Glass bottle 5mL
10062880 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10269773 Supplier Thermo Scientific Acros Supplier No. 218575000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 543-59-9 | C5H11Cl | 106.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-59-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.59
MDL Number MFCD00001015
InChI Key SQCZQTSHSZLZIQ-UHFFFAOYSA-N
Synonym pentyl chloride, pentane, 1-chloro, n-amyl chloride, n-pentyl chloride, amyl chloride, chloropentane, unii-0eg9msd3nk, 0eg9msd3nk, pentane, chloro, n-butylcarbonyl chloride
PubChem CID 10977
IUPAC Name 1-chloropentane
SMILES CCCCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60°C
Color Colorless
Density 0.8800g/mL
Boiling Point 107°C to 108°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)4Cl
Refractive Index 1.4115 to 1.4135
Quantity 500 mL
Beilstein 01,13
Merck Index 15,6
Specific Gravity 0.88
Solubility Information Solubility in water: negligible. Other solubilities: miscible with alcohol and ether
Viscosity 0.59 mPa.s (20°C)
Formula Weight 106.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloropentane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloropentane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.