missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorophthalazin-4-one, 98%

Product Code. 11401780
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401780 1 g 1g
15433707 250 mg 250mg
11411780 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11401780 Supplier Thermo Scientific Alfa Aesar Supplier No. H27515.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2257-69-4 | C8H5ClN2O | 180.59 g/mol

1-Chlorophthalazin-4-one acts as an organic and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorophthalazin-4-one acts as an organic and pharmaceutical intermediate.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2257-69-4
Molecular Formula C8H5ClN2O
Molecular Weight (g/mol) 180.59
MDL Number MFCD00102192
InChI Key QCKGMJDOJRNSMS-UHFFFAOYSA-N
Synonym 4-chlorophthalazin-1 2h-one, 4-chloro-1,2-dihydrophthalazin-1-one, 4-chlorophthalazin-1-ol, 1 2h-phthalazinone, 4-chloro, 4-chloro-1 2h-phthalazinone, 4-chlorophthalazone, 4-chloro-1-phthalazinol, 4-chlorophthalazin-1-one, 1-chlorophthalazin-4-one, 1-chlorophthalazine-4-one
PubChem CID 2775142
SMILES ClC1=NNC(=O)C2=CC=CC=C12

Specifications

Melting Point 270°C to 274°C
Quantity 1 g
IUPAC Name 4-chloro-1,2-dihydrophthalazin-1-one
Formula Weight 180.59
Percent Purity 98%
Chemical Name or Material 1-Chlorophthalazin-4-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.