missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorophthalazine, TRC

Product Code. 30135559
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135559 5 g 5g
30136130 1 g 1g
17903915 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135559 Supplier Toronto Research Chemicals Supplier No. TRCC3796605G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5784-45-2
Molecular Formula C8 H5 Cl N2
Molecular Weight (g/mol) 164.59
Synonym Phthalazine, 1-chloro-, 1-Chlorophthalazine, NSC 71104
IUPAC Name 1-chlorophthalazine
SMILES Clc1nncc2ccccc12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H5ClN2/c9-8-7-4-2-1-3-6(7)5-10-11-8/h1-5H
Formula Weight 164.0141
Percent Purity >95%
Chemical Name or Material 1-Chlorophthalazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.