missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloropropane-1,1-d2, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Chloropropane-1,1-D2 |
| CAS | 125640-23-5 |
| Molecular Formula | C3 D2 H5 Cl |
| Quantity | 0.1 g |
| Synonym | Propane-1,1-d2, 1-chloro- (9 CI), 1-Chloropropane-1,1-d2, 1-Chloropropane-1,1-D2 |
| InChI Formula | InChI=1 S/C3H7Cl/c1-2-3-4/h2-3H2,1H3/i3D2 |
| SMILES | [2 H]C([2 H])(Cl)CC |
| IUPAC Name | 1-chloro-1,1-dideuteriopropane |
| Molecular Weight (g/mol) | 80.553 |
| Formula Weight | 80.0362 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction