missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloropropane-d7, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Chloropropane-D7 |
| CAS | 761374-88-3 |
| Molecular Formula | C3 D7 Cl |
| Quantity | 0.1 g |
| Synonym | Propane-1,1,1,2,2,3,3-d7, 3-chloro- (9 CI), 3-Chloropropane-1,1,1,2,2,3,3-d7, 1-Chloropropane-d7, 1-Chloropropane-D7 |
| InChI Formula | InChI=1 S/C3H7Cl/c1-2-3-4/h2-3H2,1H3/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 1-chloro-1,1,2,2,3,3,3-heptadeuteriopropane |
| Molecular Weight (g/mol) | 85.5838 |
| Formula Weight | 85.0676 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction