missing translation for 'onlineSavingsMsg'
Learn More

1-Cyano-4-dimethylaminopyridinium tetrafluoroborate, +97%, ACROS Organics™

Product Code. 10634755 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10634755 100 mg Glass bottle 100mg
10173104 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10634755 Supplier Thermo Scientific Acros Supplier No. 397421000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 59016-56-7
Molecular Formula C8H10BF4N3
Molecular Weight (g/mol) 234.99
MDL Number MFCD00011998
InChI Key MBLVMDCQDCVKNE-UHFFFAOYSA-N
Synonym cdap, 1-cyano-4-dimethylamino pyridinium tetrafluoroborate, 1-cyano-4-dimethylamino pyridin-1-ium tetrafluoroborate, unii-p4w72066jt, 1-cyano-4-dimethylamino-pyridinium tetrafluoroborate, pubchem17735, acmc-209m9a, n-cyano-4-dimethylaminopyridinium tetrafluoroborate, 1-cyano-4-dimethylaminopyridinium tetra-fluoroborate, n-cyano-4-dimethylamino-pyridinium tetrafluoroborate
PubChem CID 9881151
IUPAC Name 1-cyano-4-(dimethylamino)pyridin-1-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C1=CC=[N+](C=C1)C#N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 196.0°C to 200.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 234.99
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Cyano-4-dimethylaminopyridinium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.