missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole, 97%

Product Code. 11398115
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398115 5 g 5g
11308125 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398115 Supplier Thermo Scientific Alfa Aesar Supplier No. L08369.06
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36140-83-7 | C8H9N3O | 163.18 g/mol

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole is used in pharma industry.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36140-83-7
Molecular Formula C8H9N3O
Molecular Weight (g/mol) 163.18
MDL Number MFCD00051954
InChI Key DDWZYWSLHBDVGR-UHFFFAOYSA-N
Synonym 1-cyanoacetyl-3,5-dimethylpyrazole, 3-3,5-dimethyl-1h-pyrazol-1-yl-3-oxopropanenitrile, 3-3,5-dimethylpyrazol-1-yl-3-oxopropanenitrile, 1-cyanoacetyl-3,5-dimethyl-1h-pyrazole, 3-3,5-dimethylpyrazolyl-3-oxopropanenitrile, cambridge id 5140887, 1h-pyrazole-1-propanenitrile, 3,5-dimethyl-, a-oxo, 1,cyanoacetyl-3,5-dimethylpyrazole, 1h-pyrazole-1-propanenitrile, 3,5-dimethyl-beta-oxo
PubChem CID 262101
IUPAC Name 3-(3,5-dimethylpyrazol-1-yl)-3-oxopropanenitrile
SMILES CC1=CC(=NN1C(=O)CC#N)C

Specifications

Melting Point 118°C to 122°C
Quantity 5 g
Beilstein 6286
Sensitivity Hygroscopic
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.