missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1,2-dicarboxylic anhydride, 97+%

Product Code. 15483478
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483478 25 g 25g
15493478 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483478 Supplier Thermo Scientific Alfa Aesar Supplier No. H56929.14
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2426-02-0 | C8H8O3 | 152.15 g/mol

It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.

Solubility
Reacts with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2426-02-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005917
InChI Key HMMBJOWWRLZEMI-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione
PubChem CID 12475
IUPAC Name 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=C1CCCC2

Specifications

Melting Point 69°C to 73°C
Quantity 25 g
UN Number UN2698
Beilstein 136335
Solubility Information Reacts with water.
Formula Weight 152.15
Percent Purity ≥97%
Chemical Name or Material 1-Cyclohexene-1,2-dicarboxylic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.