missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1-carbonitrile, 98%

Product Code. 11308345
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
11308345 2 g 2g
11318345 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 11308345

Brand: Thermo Scientific Alfa Aesar L10284.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1855-63-6 | C7H9N | 107.156 g/mol

1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1855-63-6
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00013772
InChI Key GTMWGXABXQTZRJ-UHFFFAOYSA-N
Synonym 1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm
PubChem CID 74619
IUPAC Name cyclohexene-1-carbonitrile
SMILES C1CCC(=CC1)C#N

Specifications

Density 0.947
Boiling Point 85°C to 88°C (17 mmHg)
Flash Point 83°C (181°F)
Refractive Index 1.482
Quantity 2 g
UN Number UN3276
Beilstein 385837
Solubility Information Not miscible in water.
Formula Weight 107.16
Percent Purity 98%
Chemical Name or Material 1-Cyclohexene-1-carbonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 1-Cyclohexene-1-carbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Cyclohexenecarbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.