missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexenylboronic acid pinacol ester, 97%

Product Code. 10402868 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10402868 500 mg 500mg
10309827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402868 Supplier Thermo Scientific Acros Supplier No. 430315000
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 141091-37-4 | C12H21BO2 | 208.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141091-37-4
Molecular Formula C12H21BO2
Molecular Weight (g/mol) 208.11
MDL Number MFCD05663845
InChI Key QNZFUMVTUFOLRT-UHFFFAOYSA-N
Synonym 2-cyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexen-yl-boronic acid pinacol ester, cyclohexene-1-boronic acid pinacol ester, 1-cyclohexen-1-yl-boronic acid pinacol ester, 2-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid, 2-1-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, cyclohexen-1-ylboronic acid, pinacol ester, 2-1-cyclohexen-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid pinacol ester
PubChem CID 10932675
IUPAC Name 2-(cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Tan
Boiling Point 125.0°C (0.2 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 500 mg
Formula Weight 208.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Cyclohexenylboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 1-Cyclohexenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.