missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-1-butanol, 98%, Thermo Scientific™

Product Code. 11460591
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460591 25 g 25g
11450591 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460591 Supplier Thermo Scientific Alfa Aesar Supplier No. B20090.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4352-42-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00019360
InChI Key MTUCYAOJXPTLHZ-UHFFFAOYNA-N
Synonym 1-cyclohexyl-1-butanol, cyclohexanemethanol, .alpha.-propyl, alpha-propylcyclohexanemethanol, zyklohexyl-butanol, cyclohexanemethanol, alpha-propyl, acmc-1ctfc, 1-hydroxybutyl cyclohexane, n-propyl cyclohexyl carbinol, mtucyaojxptlhz-uhfffaoysa
PubChem CID 107269
IUPAC Name 1-cyclohexylbutan-1-ol
SMILES CCCC(O)C1CCCCC1

Specifications

Density 0.902
Boiling Point 103°C to 104°C (4 mmHg)
Flash Point 108°C (228°F)
Refractive Index 1.465
Quantity 25 g
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 1-Cyclohexyl-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.