missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentene-1-carboxylic acid, 98%

Product Code. 10401284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10401284 5 g Glass bottle 5g
10396344 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10401284 Supplier Thermo Scientific Acros Supplier No. 396830050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1560-11-8 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1560-11-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001396
InChI Key PYRZPBDTPRQYKG-UHFFFAOYSA-N
Synonym 1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
PubChem CID 95964
IUPAC Name cyclopentene-1-carboxylic acid
SMILES C1CC=C(C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121.0°C to 124.0°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C5H7COOH
Quantity 5 g
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 1-Cyclopentene-1-carboxylic acid
compliance-icons
Product Identifier
  • 1-Cyclopentene-1-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.