missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentene-1-carboxylic acid, 98%

Product Code. 11400823
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11400823 1 g 1g
11410823 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11400823

Brand: Thermo Scientific Alfa Aesar A13271.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1560-11-8 | C6H8O2 | 112.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1560-11-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001396
InChI Key PYRZPBDTPRQYKG-UHFFFAOYSA-N
Synonym 1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
PubChem CID 95964
IUPAC Name cyclopentene-1-carboxylic acid
SMILES C1CC=C(C1)C(=O)O

Specifications

Melting Point 117°C to 121°C
Boiling Point 210°C
Quantity 1 g
UN Number UN3261
Beilstein 1446347
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 1-Cyclopentene-1-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.