missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenecarboxylic Acid, TRC

Product Code. 30357118
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
30357118 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 30357118

Brand: Toronto Research Chemicals TRCC98999025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclopentenecarboxylic Acid
CAS 1560-11-8
Molecular Formula C6H8O2
Quantity 25 mg
Synonym Isoaleprolic Acid, NSC 44968
InChI Formula InChI=1S/C6H8O2/c7-6(8)5-3-1-2-4-5/h3H,1-2,4H2,(H,7,8)
SMILES OC(=O)C1=CCCC1
IUPAC Name cyclopentene-1-carboxylic acid
Molecular Weight (g/mol) 112.13
Formula Weight 112.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.