missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentylethanol, 97%

Product Code. 11392156
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392156 10 g 10g
11382156 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392156 Supplier Thermo Scientific Alfa Aesar Supplier No. L03446.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 52829-98-8 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52829-98-8
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00013743
InChI Key WPCMSUSLCWXTKB-UHFFFAOYNA-N
Synonym 1-cyclopentylethan-1-ol, cyclopentanemethanol, .alpha.-methyl, alpha-methylcyclopentanemethanol, cyclopentyl ethanol, 1-hydroxyethylcyclopentane, 1-cyclopentylethanol, cyclopentanemethanol, alpha-methyl
PubChem CID 95434
IUPAC Name 1-cyclopentylethanol
SMILES CC(O)C1CCCC1

Specifications

Density 0.911
Boiling Point 68°C (8 mmHg)
Refractive Index 1.458
Quantity 10 g
Beilstein 2069851
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material 1-Cyclopentylethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.