missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentylethanol, 97%, Thermo Scientific™

Product Code. 11994641 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2 g
Unit Size:
2g
This item is not returnable. View return policy

Product Code. 11994641

Brand: Thermo Scientific Acros 441540020

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 52829-98-8
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9190g/mL
Boiling Point 169°C to 170°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C7H14O
Refractive Index 1.458
MDL Number MFCD00013743
Quantity 2 g
Specific Gravity 0.919
Synonym 1-cyclopentylethan-1-ol, cyclopentanemethanol, .alpha.-methyl, alpha-methylcyclopentanemethanol, cyclopentyl ethanol, 1-hydroxyethylcyclopentane, 1-cyclopentylethanol, cyclopentanemethanol, alpha-methyl
InChI Key WPCMSUSLCWXTKB-UHFFFAOYNA-N
SMILES CC(O)C1CCCC1
IUPAC Name 1-cyclopentylethanol
Molecular Weight (g/mol) 114.19
PubChem CID 95434
Formula Weight 114.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Cyclopentylethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.