missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonyl chloride, 98%

Product Code. 11326936
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11326936 10 g 10g
11336936 50 g 50g
2 options
This item is not returnable. View return policy

Product Code. 11326936

Brand: Thermo Scientific Alfa Aesar L06642.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 61652-81-1 | C10H21ClO2S | 240.79 g/mol

1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61652-81-1
Molecular Formula C10H21ClO2S
Molecular Weight (g/mol) 240.79
MDL Number MFCD00051746
InChI Key WCKKWABZCCKDTN-UHFFFAOYSA-N
Synonym 1-decanesulfonyl chloride, decanesulfonylchloride, decanesulfonyl chloride, decylsulfonyl chloride, 1-decanesulphonylchloride, acmc-1b9lu
PubChem CID 109107
IUPAC Name decane-1-sulfonyl chloride
SMILES CCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 30°C to 34°C
Quantity 10 g
UN Number UN3261
Beilstein 1771852
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 240.79
Percent Purity 98%
Chemical Name or Material 1-Decanesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.