missing translation for 'onlineSavingsMsg'
Learn More

1-Decanol, 98+%

Product Code. 11453517
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453517 2500 mL 2500mL
11443517 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453517 Supplier Thermo Scientific Alfa Aesar Supplier No. A17288.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-30-1 | C10H22O | 158.285 g/mol

1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.

Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-30-1
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00004747
InChI Key MWKFXSUHUHTGQN-UHFFFAOYSA-N
Synonym 1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
PubChem CID 8174
ChEBI CHEBI:28903
IUPAC Name decan-1-ol
SMILES CCCCCCCCCCO

Specifications

Melting Point 6°C
Density 0.828
Boiling Point 230°C to 232°C
Flash Point 82°C (179°F)
Assay Percent Range >98%
Odor Sweet
Linear Formula CH3(CH2)8CH2OH
Refractive Index 1.437
Quantity 2500 mL
UN Number UN3082
Beilstein 1735221
Merck Index 14,2855
Solubility Information Soluble in alcohol,ether; Insoluble in water;Miscible with carbon tetrachloride,ethanol,ether,acetone,benzene,chloroform and glacial acetic acid; Immiscible with water.
Formula Weight 158.29
Percent Purity ≥98%
Chemical Name or Material 1-Decanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Decanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Decanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.