missing translation for 'onlineSavingsMsg'
Learn More

1-Decanol, TRC

Product Code. 30349124
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349124 Supplier Toronto Research Chemicals Supplier No. TRCD21254025G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Decanol
CAS 112-30-1
Molecular Formula C10 H22 O
Quantity 25 g
Synonym Decyl alcohol (8CI), 1-Hydroxydecane, Alfol 10, Antak, Capric alcohol, Caprinic alcohol, Capryl alcohol, Conol 1098, Conol 10N, Decanol, Epal 10, Kalcohl 10, Kalcohl 1098, Kalcohl 10H, Kalcol 1098, Lorol C 10-98, NSC 406313, Nacol 10, Nacol 10-99, Nafol 10, Nafol 10D, Nonylcarbinol, Royaltac, Sipol L 10, T 148, n-Decanol, n-Decyl alcohol
InChI Formula InChI=1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3
SMILES CCCCCCCCCCO
IUPAC Name decan-1-ol
Molecular Weight (g/mol) 158.28
Formula Weight 158.1671
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.