missing translation for 'onlineSavingsMsg'
Learn More

1-Decene, 94%

Product Code. 11356996
Change view
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356996 1000 mL 1 L
11346996 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356996 Supplier Thermo Scientific Alfa Aesar Supplier No. L06800.AU
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872-05-9 | C10H20 | 140.27 g/mol

1-Decene exists as an alpha olefin, it is used as a monomer in copolymers. It is used as an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decene exists as an alpha olefin, it is used as a monomer in copolymers. It is used as an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics.

Solubility
Miscible in Ether, Ethanol. Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-05-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009577
InChI Key AFFLGGQVNFXPEV-UHFFFAOYSA-N
Synonym 1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
PubChem CID 13381
ChEBI CHEBI:87315
IUPAC Name dec-1-ene
SMILES CCCCCCCCC=C

Specifications

Melting Point -66°C
Boiling Point 170°C to 171°C
Flash Point 49°C (120°F)
Linear Formula CH3(CH2)7CH=CH2
Quantity 1000 mL
UN Number UN3295
Beilstein 1737236
Formula Weight 140.27
Percent Purity 94%
Chemical Name or Material 1-Decene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.