missing translation for 'onlineSavingsMsg'
Learn More

1-Decene, 96%, remainder isomers

Product Code. 11397428
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397428 2 kg 2kg
11307438 100 g 100g
11317438 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397428 Supplier Thermo Scientific Alfa Aesar Supplier No. 031213.A3
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872-05-9 | C10H20 | 140.27 g/mol

1-Decene is the only isomer of industrial importance. it is used as a monomer in copolymers and is an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics. 1-Decene has been isolated from the leaves and rhizome of the plant Farfugium japonicum and has been detected as the initial product in the microbial degradation of n-decane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-05-9
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00009577
InChI Key AFFLGGQVNFXPEV-UHFFFAOYSA-N
Synonym 1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
PubChem CID 13381
ChEBI CHEBI:87315
IUPAC Name dec-1-ene
SMILES CCCCCCCCC=C

Specifications

Melting Point -66°C
Color Colorless
Density 0.741 g/mL
Boiling Point 170°C to 171°C
Flash Point 49°C (120°F)
Assay Percent Range 96%
Linear Formula CH3(CH2)7CH=CH2
Refractive Index 1.421
Quantity 2 kg
UN Number UN3295
Beilstein 1737236
Solubility Information Alkenes are virtually insoluble in water,but dissolve in organic solvents.
Formula Weight 140.27
Percent Purity 96%
Physical Form Liquid
Assay Remainder isomers
Chemical Name or Material 1-Decene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.