missing translation for 'onlineSavingsMsg'
Learn More

1-Decene, TRC

Product Code. 30368640
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368640 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368640 Supplier Toronto Research Chemicals Supplier No. TRCD2255551G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 872-05-9
Molecular Formula C10 H20
Quantity 1 g
Synonym 1-n-Decene, Dialen 10, Gulftene 10, Linealene 10, NSC 62122, Neodene 10, n-1-Decene, α-Decene
InChI Formula InChI=1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3
SMILES CCCCCCCCC=C
IUPAC Name dec-1-ene
Molecular Weight (g/mol) 140.27
Formula Weight 140.1565
Chemical Name or Material 1-Decene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.