missing translation for 'onlineSavingsMsg'
Learn More

1-Decylboronic acid, 98%

Product Code. 11394326
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394326 1 g 1g
11304336 5 g 5g
11314336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394326 Supplier Thermo Scientific Alfa Aesar Supplier No. L19957.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24464-63-9 | C10H23BO2 | 186.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24464-63-9
Molecular Formula C10H23BO2
Molecular Weight (g/mol) 186.102
MDL Number MFCD01074638
InChI Key TUTSEMMBKGBUGD-UHFFFAOYSA-N
Synonym n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d
PubChem CID 4186635
IUPAC Name decylboronic acid
SMILES B(CCCCCCCCCC)(O)O

Specifications

Melting Point 86°C to 89°C
Quantity 1 g
Beilstein 1743934
Formula Weight 186.1
Percent Purity 98%
Chemical Name or Material 1-Decylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.