missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxy-1-nitro-L-iditol hemihydrate, 99%, Thermo Scientific™

Product Code. 10334352 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
10334352 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 10334352

Brand: Thermo Scientific Acros 353721000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 207226-23-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 89°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H28N2O15
MDL Number MFCD00191972
Quantity 100 mg
Specific Rotation -4.5
Synonym 1-deoxy-1-nitro-l-iditol hemihydrate, 2s,3r,4r,5s-6-nitrohexane-1,2,3,4,5-pentol hydrate
InChI Key HQAUHMYCAWVBKQ-UHFFFAOYNA-N
SMILES O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O
IUPAC Name (2S,3R,4R,5S)-6-nitrohexane-1,2,3,4,5-pentol;hydrate
Molecular Weight (g/mol) 440.36
PubChem CID 73995060
Formula Weight 220.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1-Deoxy-1-nitro-L-iditol hemihydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.