missing translation for 'onlineSavingsMsg'
Learn More

1-Desmethyl 2-Methyl Granisetron (Granisetron Impurity A), TRC

Product Code. 17964075
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964075 50 mg 50mg
17923765 5 mg 5mg
17911335 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964075 Supplier Toronto Research Chemicals Supplier No. TRCD29208550MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127472-42-8
Molecular Formula C18 H24 N4 O
Molecular Weight (g/mol) 312.41
Synonym 2H-Indazole-3-carboxamide, 2-methyl-N-(9-methyl-9-azabicyclo[3.3.1]non-3-yl)-, endo- (9CI), 9-Azabicyclo[3.3.1]nonane, 2H-indazole-3-carboxamide deriv., 2-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide, 2-methyl-N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide
IUPAC Name 2-methyl-N-[(1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]indazole-3-carboxamide
SMILES CN1[C@@H]2CCC[C@H]1C[C@H](C2)NC(=O)c3c4ccccc4nn3C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H24N4O/c1-21-13-6-5-7-14(21)11-12(10-13)19-18(23)17-15-8-3-4-9-16(15)20-22(17)2/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23)/t12-,13+,14-
Formula Weight 312.195
Percent Purity >95%
Chemical Name or Material 2-methyl-N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-2H-indazole-3-carboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.