missing translation for 'onlineSavingsMsg'
Learn More

1-Desmethyl Granisetron (Granisetron Impurity B), TRC

Product Code. 30136587
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136587 5 mg 5mg
30134858 1 mg 1mg
30012481 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136587 Supplier Toronto Research Chemicals Supplier No. TRCD2920385MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107007-95-4
Molecular Formula C17 H22 N4 O
Molecular Weight (g/mol) 298.38
Synonym Granisetron Hydrochloride Imp. B (EP), N-[(1R,3r,5S)-9-Methyl-9-azabicyclo-[3.3.1]non-3-yl]-1H-indazole-3-carboxamide, N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-Indazole-3-carboxamide, endo-N-(9-Methyl-9-azabicyclo[3.3.1]non-3-yl)-1H-Indazole-3-carboxamide, 9-Azabicyclo[3.3.1]nonane, 1H-indazole-3-carboxamide deriv.
IUPAC Name N-[(1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-yl]-1H-indazole-3-carboxamide
SMILES CN1[C@@H]2CCC[C@H]1C[C@H](C2)NC(=O)c3n[nH]c4ccccc34

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H22N4O/c1-21-12-5-4-6-13(21)10-11(9-12)18-17(22)16-14-7-2-3-8-15(14)19-20-16/h2-3,7-8,11-13H,4-6,9-10H2,1H3,(H,18,22)(H,19,20)/t11-,12+,13-
Formula Weight 298.1794
Percent Purity >95%
Chemical Name or Material N-[(1R,3r,5S)-9-Methyl-9-azabicyclo-[3.3.1]non-3-yl]-1H-indazole-3-carboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.