missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-2-propanol (Dimepranol), TRC

Product Code. 17981065
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981065 1 g 1g
17914495 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17981065 Supplier Toronto Research Chemicals Supplier No. TRCD4604001G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-16-7
Molecular Formula C5 H13 N O
Molecular Weight (g/mol) 103.16
Synonym 1-(Dimethylamino)-2-propanol, (±)-1-(Dimethylamino)-2-propanol, (±)-1-(N,N-Dimethylamino)-2-propanol, 1-Methyl-2-(dimethylamino)ethanol, 2-(Dimethylamino)-1-methyl-1-ethanol, 2-(Dimethylamino)-1-methylethanol, 3-(Dimethylamino)-2-propanol, Bisomer Amine D 700, DL-1-(Dimethylamino)-2-propanol, DMA 2P, Dimepranol, Dimethyl(2-hydroxypropyl)amine, Dimethylisopropanolamine, N,N-Dimethyl-2-hydroxy-1-propanamine, N,N-Dimethyl-2-hydroxypropylamine, N,N-Dimethylamino-2-propanol, N,N-Dimethylisopropanolamine, NSC 3163
IUPAC Name 1-(dimethylamino)propan-2-ol
SMILES CC(O)CN(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3
Formula Weight 103.0997
Chemical Name or Material Dimepranol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.