missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-2-propyne, 98%

Product Code. 11338745
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338745 25 g 25g
11328745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338745 Supplier Thermo Scientific Alfa Aesar Supplier No. L12336.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7223-38-3 | C5H9N | 83.134 g/mol

1-Dimethylamino-2-propyne is used in the synthesis of core shell systems based on noble metal nanoparticles coated by copolymers (polyacetylenes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dimethylamino-2-propyne is used in the synthesis of core shell systems based on noble metal nanoparticles coated by copolymers (polyacetylenes).

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to carbon dioxide. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7223-38-3
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00008575
InChI Key ILBIXZPOMJFOJP-UHFFFAOYSA-N
Synonym 3-dimethylamino-1-propyne, 1-dimethylamino-2-propyne, 2-propyn-1-amine, n,n-dimethyl, n,n-dimethylpropargylamine, dimethyl prop-2-yn-1-yl amine, n,n-dimethyl-2-propyn-1-amine, dimethyl prop-2-ynyl amine, n,n-dimethyl propargylamine, n,n-dimethyl-2-propynylamine, dimethylpropargylamine
PubChem CID 81643
IUPAC Name N,N-dimethylprop-2-yn-1-amine
SMILES CN(C)CC#C

Specifications

Density 0.772
Boiling Point 80°C to 81°C
Flash Point −7°C (19°F)
Refractive Index 1.42
Quantity 25 g
UN Number UN2733
Beilstein 1071222
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 83.13
Percent Purity 98%
Chemical Name or Material 1-Dimethylamino-2-propyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.