missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanethiol, TRC

Product Code. 30379508
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30379508 500 mg -
30371245 250 mg -
30378113 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30379508 Supplier Toronto Research Chemicals Supplier No. TRCD678868500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7773-83-3
Molecular Formula C22H46S
Molecular Weight (g/mol) 342.67
Synonym Docosanethiol, Docosyl Mercaptan, NSC 97595, n-Docosanethiol
IUPAC Name docosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCCCS

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H46S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23/h23H,2-22H2,1H3
Formula Weight 342.332
Chemical Name or Material 1-Docosanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.