missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

Product Code. 10288033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10288033 5 g Glass bottle 5g
10782244 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10288033 Supplier Thermo Scientific Acros Supplier No. 396720050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2386-53-0 | C12H25NaO3S | 272.379 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-53-0
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
PubChem CID 23665726
IUPAC Name sodium;dodecane-1-sulfonate
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Absorbance 0.02 max. (C=0.18 M in hot water) at 230nm, 0.03 max. (C=0.18 M in hot water) at 220nm, 0.05 max. (C=0.18 M in hot water) at 210nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C12H25NaO3S
Linear Formula CH3(CH2)11SO3Na
Quantity 5 g
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 272.38
Formula Weight 272.38
Percent Purity 99+%
Grade Ion Pair Chromatography
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecanesulfonic acid, sodium salt, Anhydrous, Ion pair chromatography
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.