missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonic acid, sodium salt, 99+%

Product Code. 10786181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10786181 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10786181 Supplier Thermo Scientific Acros Supplier No. 147900010
  • ONLINE EXCLUSIVE PRICE
    30.90 €
    30.00 € / 1g
    Save 0.90 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2386-53-0 | C12H25NaO3S | 272.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2386-53-0
CAS Min % 99.0
CAS Max % 100.0
Melting Point >300.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C12H25NaO3S
Linear Formula CH3(CH2)11SO3Na
MDL Number MFCD00007527
Quantity 1 g
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
Solubility Information Solubility in water: soluble.
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;dodecane-1-sulfonate
Molecular Weight (g/mol) 272.38
PubChem CID 23665726
Formula Weight 272.38
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 1-Dodecanesulfonic acid, sodium salt, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.