missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonic Acid Sodium Salt, Ion Pair Chromatography, Fisher Chemical

Product Code. 10659803 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Plastic powder jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10659803 5 g Plastic powder jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10659803 Supplier Fisher Chemical Supplier No. D/5297/44
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 300°C
CAS 2386-53-0
Color White
Physical Form Solid
Quantity 5 g
Molecular Formula C12H25NaO3S
MDL Number 7527
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;dodecane-1-sulfonate
Molecular Weight (g/mol) 272.379
PubChem CID 23665726
Formula Weight 272.38g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.