missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanesulfonyl chloride, 97%

Product Code. 11364514
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364514 25 g 25g
11354514 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364514 Supplier Thermo Scientific Alfa Aesar Supplier No. L01224.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10147-40-7 | C12H25ClO2S | 268.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10147-40-7
Molecular Formula C12H25ClO2S
Molecular Weight (g/mol) 268.84
MDL Number MFCD00014728
InChI Key PBULHKIPTBIZHO-UHFFFAOYSA-N
Synonym 1-dodecanesulfonyl chloride, 1-dodecanesulfonylchloride, unii-g45gs200po, 1-dodecanesulphonyl chloride, dodecylsulfonyl chloride, dodecane-1-sulphonyl chloride, chlorododecylsulfone, dodecanesulfonylchloride, acmc-20a5qf, dodecanesulfonyl chloride
PubChem CID 66280
IUPAC Name dodecane-1-sulfonyl chloride
SMILES CCCCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 39°C to 42°C
Boiling Point 126°C to 130°C (4 mmHg)
Linear Formula CH3(CH2)11SO2Cl
Quantity 25 g
UN Number UN3261
Beilstein 1778092
Sensitivity Moisture sensitive
Formula Weight 268.84
Percent Purity ≥98%
Chemical Name or Material 1-Dodecanesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.