missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanethiol, 98%, Thermo Scientific Chemicals

Product Code. 10273950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
10 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10273950 500 mL Glass bottle 500mL
10358743 2.5 L Glass bottle 2.5 L
11965721 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10273950 Supplier Thermo Scientific Acros Supplier No. 117625000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-55-0
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00004885
InChI Key WNAHIZMDSQCWRP-UHFFFAOYSA-N
Synonym 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
PubChem CID 8195
IUPAC Name dodecane-1-thiol
SMILES CCCCCCCCCCCCS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -7.0°C
Density 0.8400g/mL
Boiling Point 266.0°C to 283.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11SH
Refractive Index 1.4580 to 1.4600
Quantity 500 mL
Fieser 16,356
Specific Gravity 0.84
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, slightly soluble in alcohol
Formula Weight 202.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dodecanethiol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Dodecanethiol
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.