missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanethiol, TRC

Product Code. 30369905
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369905 50 mL -
30379477 5 mL 5mL
30374751 10 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369905 Supplier Toronto Research Chemicals Supplier No. TRCD49479350ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-55-0
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.4
Synonym 1-Dodecyl Mercaptan, 1-Dodecylthiol, 1-Mercaptododecane, Lauryl Mercaptan, NDM, NSC 814, Thiokalcol 20, n-Dodecanethiol, n-Dodecyl Mercaptan, n-Dodecyl Thiolate, n-Dodecylthiol, n-Lauryl Mercaptan
SMILES CCCCCCCCCCCCS

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C12H26S/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3
Formula Weight 202.1755
Percent Purity >95%
Chemical Name or Material 1-Dodecanethiol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.