missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol, Reagent grade, 98%, Honeywell™

Product Code. 15691180 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Poly Bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15691180 1 L Poly Bottle 1 L
15611190 250 mL Poly Bottle 250mL
15601190 2.5 L Poly Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15691180 Supplier Honeywell Research Chemicals Supplier No. 1267991L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Melting Point 22°C to 26°C
Color Colorless
Density 0.833g/mL at 25°C
Boiling Point 260°C to 262°C
Flash Point 121°C (249.8°F)
Packaging Poly Bottle
Linear Formula CH3(CH2)11OH
Refractive Index n20/D 1.442(lit.)
Quantity 1 L
UN Number UN3077
Beilstein 1738860
Solubility Information Slightly soluble in water, 1g/L at 23°C
Vapor Density 7.4 (vs air)
Formula Weight 186.33g/mol
Percent Purity 98%
Grade Reagent
Chemical Name or Material 1-Dodecanol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.