missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecene, 93-95%, Thermo Scientific Chemicals

Catalog Number 10071070
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Plastic drum
This item is not returnable. View return policy
112-41-4
C12H24
168.32
MFCD00008961
CRSBERNSMYQZNG-UHFFFAOYSA-N
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
8183
dodec-1-ene
CCCCCCCCCCC=C
This item is not returnable. View return policy
112-41-4
C12H24
168.32
MFCD00008961
CRSBERNSMYQZNG-UHFFFAOYSA-N
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
8183
dodec-1-ene
CCCCCCCCCCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 93.0
CAS Max % 100.0
Melting Point -35.0°C
Density 0.7500g/mL
Boiling Point 213.0°C
Flash Point 77°C
Assay Percent Range 93% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9CH=CH2
Refractive Index 1.4284 to 1.4304
Quantity 100 g
Beilstein 01,225
Specific Gravity 0.75
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 168.32
Percent Purity 93 to 95%
Chemical Name or Material 1-Dodecene, 93-95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'