missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyl isocyanate, 98%

Product Code. 11348086
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348086 1 g 1g
11353017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348086 Supplier Thermo Scientific Alfa Aesar Supplier No. L13380.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4202-38-4 | C13H25NO | 211.349 g/mol

1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4202-38-4
Molecular Formula C13H25NO
Molecular Weight (g/mol) 211.349
MDL Number MFCD00041744
InChI Key YIDSTEJLDQMWBR-UHFFFAOYSA-N
Synonym dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, isocyanic acid dodecyl ester
PubChem CID 77873
IUPAC Name 1-isocyanatododecane
SMILES CCCCCCCCCCCCN=C=O

Specifications

Density 0.877
Boiling Point 157°C to 161°C (21 mmHg)
Flash Point 134°C (273°F)
Odor Irritating
Refractive Index 1.441
Quantity 1 g
UN Number UN2206
Beilstein 1774823
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 211.35
Percent Purity 98%
Chemical Name or Material 1-Dodecyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.