missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, 97%

Product Code. 11347915
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347915 25 g 25g
11337915 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347915 Supplier Thermo Scientific Alfa Aesar Supplier No. L02977.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 765-03-7 | C12H22 | 166.31 g/mol

1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-03-7
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00008960
InChI Key ZVDBUOGYYYNMQI-UHFFFAOYSA-N
Synonym 1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
PubChem CID 69821
IUPAC Name dodec-1-yne
SMILES CCCCCCCCCCC#C

Specifications

Melting Point -19°C
Density 0.779
Boiling Point 214°C to 216°C
Flash Point 79°C (174°F)
Refractive Index 1.434
Quantity 25 g
Beilstein 1741320
Solubility Information Not miscible in water.
Formula Weight 166.31
Percent Purity 97%
Chemical Name or Material 1-Dodecyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.