missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 1-EBIO
GREENER_CHOICE

Product Code. 30001173
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001173 50 mg 50mg
30001172 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001173 Supplier Tocris Bioscience™ Supplier No. 1041/50
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Activator of epithelial KCa channels

Activator of epithelial KCa channels, stimulates a large and sustained trans-epithelial Cl- secretory response across T84 monolayers. Induces hyperpolarization to the same magnitude as ACh in aortic value endothelial cells. Promotes embryonic stem cell (ESC) differentiation into cardiomyocytes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10045-45-1
Molecular Formula C9H10N2O

Specifications

Quantity 50 mg
Formula Weight 162.19
Purity >99%
Molecular Weight (g/mol) 162.19
Chemical Name or Material 1-EBIO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.