missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol, 90+%, remainder 2-ethoxy-1-propanol

Product Code. 11480560
Click to view available options
Quantity:
500 mL
4 x 500 mL
Unit Size:
2000mL
500mL
This item is not returnable. View return policy

Product Code. 11480560

Brand: Thermo Scientific Alfa Aesar 043306.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol

1-Ethoxy-2-propanol is used as a solvent for a wide variety of resins such as epoxies, acrylic, alkyds, polyesters, nitrocellulose, and polyurethanes. It is also used in the surface coating and printing industries, thereby it regulate the flow, levelling, and coalescence of both surface coatings. It acts as an intermediate in the production of agro-chemicals and de-icing, and anti-icing formulations. It finds application as to make cleaning products, as grease and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1569-02-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00067050
InChI Key JOLQKTGDSGKSKJ-UHFFFAOYNA-N
Synonym 1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
PubChem CID 15287
IUPAC Name 1-ethoxypropan-2-ol
SMILES CCOCC(C)O

Specifications

Melting Point -100°C
Color Colorless
Density 0.896 g/mL
Boiling Point 132°C
Flash Point 42°C (108°F)
Linear Formula CH3CH2OCH2CH(OH)CH3
Refractive Index 1.4065
Quantity 500 mL
UN Number UN1987
Solubility Information Miscible with water.
Formula Weight 104.15
Percent Purity ≥90%
Physical Form Liquid
Assay Remainder 2-ethoxy-1-propanol
Chemical Name or Material 1-Ethoxy-2-propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethoxy-2-propanol, 90+%, remainder 2-ethoxy-1-propanol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : 1-Ethoxy-2-propanol,2-Ethoxypropanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.