missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxycarbonyl-4-piperidone, 98+%

Product Code. 11433794
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433794 250 g 250g
11413794 10 g 10g
11423794 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433794 Supplier Thermo Scientific Alfa Aesar Supplier No. A11833.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 29976-53-2 | C8H13NO3 | 171.196 g/mol

1-Ethoxycarbonyl-4-piperidone is used in the preparation of 3-hydrazinopyridazines as antihypertensive agents as well as γ-carboline derivatives as potential serotonergic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethoxycarbonyl-4-piperidone is used in the preparation of 3-hydrazinopyridazines as antihypertensive agents as well as γ-carboline derivatives as potential serotonergic agents.

Solubility
Soluble in water.500 g/L at 20°C

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Protect from heat. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29976-53-2
Molecular Formula C8H13NO3
Molecular Weight (g/mol) 171.196
MDL Number MFCD00006188
InChI Key LUBGFMZTGFXIIN-UHFFFAOYSA-N
Synonym n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
PubChem CID 34767
IUPAC Name ethyl 4-oxopiperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(=O)CC1

Specifications

Density 1.135
Boiling Point 95°C to 98°C (1 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.475
Quantity 250 g
Beilstein 640967
Solubility Information Soluble in water.500g/L at 20°C
Formula Weight 171.2
Percent Purity 99%
Chemical Name or Material 1-Ethoxycarbonyl-4-piperidone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.