missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific Chemicals

Product Code. 10295980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295980 25 g Plastic bottle 25g
10012180 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295980 Supplier Thermo Scientific Acros Supplier No. 170290250
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5417-82-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00001852
InChI Key BOSVWXDDFBSSIZ-UHFFFAOYSA-N
Synonym 1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene propanedinitrile, propanedinitrile, 1-ethoxyethylidene, 1-ethoxyethylidenemalononitrile, 1-ethoxyethylidene propanedinitrile, 2-1-ethoxyethylidene-propanedinitrile, propanedinitrile, 2-1-ethoxyethylidene, ethoxyethylidene methane-1,1-dicarbonitrile, malononitrile, 1-ethoxyethylidene-6ci,7ci,8ci
PubChem CID 79450
IUPAC Name 2-(1-ethoxyethylidene)propanedinitrile
SMILES CCOC(=C(C#N)C#N)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 89.0°C to 93.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(OC2H5)=C(CN)2
Quantity 25 g
Formula Weight 136.15
Percent Purity 99+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material (1-Ethoxyethylidene)malononitrile, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • (1-Ethoxyethylidene)malononitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.