missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one, Thermo Scientific™

Product Code. 17525535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17525535 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17525535 Supplier Thermo Scientific Acros Supplier No. 186310010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

Alpha Vector ETHYLDIHYDROBENZIMIDAZOLONE
CAS 10045-45-1
Melting Point 120°C to 126°C
Color White to beige
Infrared Spectrum Conforms
Packaging Glass Bottle
Molecular Formula C9H10N2O
MDL Number MFCD00005715
Quantity 1 g
InChI Key CXUCKELNYMZTRT-UHFFFAOYSA-N
SMILES CCN1C(=O)NC2=CC=CC=C12
IUPAC Name 1-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Molecular Weight (g/mol) 162.19
Physical Form Powder
Chemical Name or Material 1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.