missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethyl-1H-1,2,4-triazol-5-yl)methanol, 97%, Thermo Scientific™

Product Code. 10603195
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10603195 1 g Amber Glass Bottle 1g
10235894 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10603195 Supplier Thermo Scientific Maybridge Supplier No. MO08364DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 215868-81-8
Molecular Formula C5H9N3O
Molecular Weight (g/mol) 127.15
MDL Number MFCD09966168
InChI Key ZQYXCGFXZPLEKX-UHFFFAOYSA-N
Synonym 1-ethyl-1h-1,2,4-triazol-5-yl methanol, 2-ethyl-1,2,4-triazol-3-yl methanol, 2-ethyl-2h-1,2,4-triazol-3-yl methanol, 1h-1,2,4-triazole-5-methanol, 1-ethyl, 2-ethyl-2h-1,2,4 triazol-3-yl methanol, 1h-1,2,4-triazole-5-methanol,1-ethyl, 1-ethyl-5-hydroxymethyl-1,2,4-triazole, 1-ethyl-1,2,4-triazol-5-yl methan-1-ol, 2-ethyl-2h-1,2,4 triazol-3-yl-methanol
PubChem CID 11412385
SMILES CCN1N=CN=C1CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name (1-ethyl-1H-1,2,4-triazol-5-yl)methanol
Formula Weight 127.15
Percent Purity 97%
Chemical Name or Material (1-Ethyl-1H-1, 2, 4-triazol-5-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.