missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-azetidinecarboxylic Acid, TRC

Product Code. 30180194
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180194 10 mg 10mg
30179957 5 mg 5mg
30180133 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180194 Supplier Toronto Research Chemicals Supplier No. TRCA96556810MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106887-13-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
IUPAC Name 1-ethylazetidine-3-carboxylic acid
SMILES CCN1CC(C1)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H11NO2/c1-2-7-3-5(4-7)6(8)9/h5H,2-4H2,1H3,(H,8,9)
Formula Weight 129.079
Chemical Name or Material 1-Ethyl-3-azetidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.