missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium acetate, 97%

Product Code. 15485538
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485538 50 g 50g
15475538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485538 Supplier Thermo Scientific Alfa Aesar Supplier No. H59277.18
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 143314-17-4 | C8H14N2O2 | 170.212 g/mol

It is used as a solvent and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent and as pharmaceutical intermediate.

Solubility
Soluble in water, acetonitrile, acetone, dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143314-17-4
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.212
MDL Number MFCD06798186
InChI Key XIYUIMLQTKODPS-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium acetate, basionic tm bc 01, emim ac, 1-ethyl-3-methylimidazoliumacetate, basionics™ bc 01, acmc-1c26d, 1h-imidazolium, 3-ethyl-1-methyl-, acetate 1:1, 1-methyl-3-ethylimidazolium acetate, 1-ethyl-3-methylimidazolium acetate hplc
PubChem CID 11658353
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;acetate
SMILES CCN1C=C[N+](=C1)C.CC(=O)[O-]

Specifications

Melting Point -20°C
Density 1.027 g/mL
Flash Point 164°C (327°F)
Refractive Index n20/D 1.502
Quantity 50 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water,acetonitrile,acetone,dichloromethane.
Formula Weight 170.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.